Methyl 3-chloroisonicotinate 97%

Supplier: Ambeed
Warning

A227960-1G A227960-5G A227960-10G
76722-180EA 19.1 USD
76722-180 76722-182 77667-894
Methyl 3-chloroisonicotinate 97%
Methyl 3-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
MDL Number: MFCD09910337
CAS Number: 98273-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR