trans-2-Amino-cyclohexanol 96%

Supplier: Ambeed
Danger

Synonyms: trans-2-Aminocyclohexanol

A227700-1G A227700-25G A227700-5G A227700-100G
76722-174EA 6 USD
76722-174 76722-176 76722-178 77872-160
trans-2-Amino-cyclohexanol 96%
trans-2-Amino-cyclohexanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.174 g/mol
MDL Number: MFCD03427291
CAS Number: 6982-39-4
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR