Tris(1-Chloro-2-Propyl) Phosphate 98% (mixture of isomers)

Supplier: Ambeed
Warning

A226925-100G A226925-1000G A226925-500G
76722-140EA 32 USD
76722-140 77667-884 77667-886
Tris(1-Chloro-2-Propyl) Phosphate 98% (mixture of isomers)
Tris(1-Chloro-2-Propyl) Phosphate

Precaución: for Research Use Only

Formula: C₉H₁₈Cl₃O₄P
MW: 327.56 g/mol
MDL Number: MFCD00040408
CAS Number: 13674-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR