3-(N-Morpholino)propanesulfonic acid, hemisodium salt 97%

Supplier: Ambeed
Warning

Synonyms: 3-Morpholinopropane-1-sulphonic acid, sodium salt(2:1), MOPS hemisodium salt

A225387-25G A225387-500G
76722-100EA 9.55 USD
76722-100 76722-102
3-(N-Morpholino)propanesulfonic acid, hemisodium salt 97%
3-(N-Morpholino)propanesulphonic acid, hemisodium salt

Precaución: for Research Use Only

Formula: C₁₄H₃₀N₂NaO₈S₂
MW: 441.5200 g/mol
MDL Number: MFCD00079461
CAS Number: 117961-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR