Phenothiazine 98%

Supplier: Ambeed
Danger

Synonyms: 10H-Phenothiazine

A224816-100G A224816-500G
76722-078EA 5.31 USD
76722-078 77568-174 77600-804
Phenothiazine 98%
Phenothiazine

Precaución: For research use only.

Formula: C₁₂H₉NS
MDL Number: MFCD00005015
CAS Number: 92-84-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR