Methyl8-bromooctanoate 98%

Supplier: Ambeed
Warning

A224536-100MG A224536-250MG A224536-5G A224536-1G
76722-068EA 81.69 USD
76722-068 76722-070 76722-072 77593-462
Methyl8-bromooctanoate 98%
Methyl8-bromooctanoate

Precaución: for Research Use Only

Formula: C₉H₁₇BrO₂
MW: 237.13 g/mol
MDL Number: MFCD02258673
CAS Number: 26825-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR