N-Me-Leu-Obzl P-Tosylate 98%

Supplier: Ambeed
Warning

Synonyms: N-Me-Leu-OBzl·p-tosylate, H-N-Me-Leu-OBzl·TosOH

A224426-1G A224426-250MG
76722-056EA 40 USD
76722-056 76722-058
N-Me-Leu-Obzl P-Tosylate 98%
N-Me-Leu-Obzl P-Tosylate

Precaución: for Research Use Only

Formula: C₂₁H₂₉NO₅S
MDL Number: MFCD00083659
CAS Number: 42807-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR