Methyl-4-bromo-3-hydroxybenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-bromo-3-hydroxybenzoate
A224401-100G
A224401-10G
A224401-1G
A224401-250MG
A224401-25G
A224401-5G
76722-044EA
507.12
USD
76722-044
76722-046
76722-048
76722-050
76722-052
76722-054
Methyl-4-bromo-3-hydroxybenzoate 97%
Methyl 4-bromo-3-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₇BrO₃ MW: 231.0400 g/mol |
MDL Number:
MFCD08056372 CAS Number: 106291-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...