Silver phosphate 99%

Supplier: Ambeed
Danger

A224116-25G A224116-5G A224116-100G
76722-038EA 33.95 USD
76722-038 76722-040 76722-042 77872-141
Silver phosphate 99%
Silver phosphate

Precaución: for Research Use Only

Formula: Ag₃O₄P
MDL Number: MFCD00014148
CAS Number: 7784-09-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR