Sorbitan monopalmitate Saponification Value 145-155

Supplier: Ambeed
Warning

A223073-1000G A223073-100G A223073-500G
76722-016EA 22 USD
76722-016 76722-018 76722-020
Sorbitan monopalmitate Saponification Value 145-155
Sorbitan monopalmitate

Precaución: for Research Use Only

Formula: C₂₂H₄₂O₆
MDL Number: MFCD00080946
CAS Number: 26266-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR