Raltitrexed 98%

Supplier: Ambeed
Warning

Synonyms: N-[[5-[[(1,4-Dihydro-2-methyl-4-oxo-6-quinazolinyl)methyl]methylamino]-2-thienyl]carbonyl]-L-glutamic acid, (S)-2-(5-(Methyl((2-methyl-4-oxo-1,4-dihydroquinazolin-6-yl)methyl)amino)thiophene-2-carboxamido)pentanedioic acid, ICI-D-1694, ZD-1694

A222261-10MG A222261-1MG A222261-250MG A222261-50MG A222261-1G A222261-100MG
76721-998EA 38.2 USD
76721-998 76722-000 76722-002 76722-004 77568-142 77667-848
Raltitrexed 98%
Raltitrexed

Precaución: For research use only.

Formula: C₂₁H₂₂N₄O₆S
MW: 458.49 g/mol
MDL Number: MFCD00864168
CAS Number: 112887-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR