H-Phe(4-Br)-OMe·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Phe(4-Br)-Ome.HCl

A221894-1G A221894-5G
76721-990EA 22.28 USD
76721-990 77593-436
H-Phe(4-Br)-OMe·HCl 97%
H-Phe(4-Br)-OMe·HCl

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂·HCl
MW: 294.5 g/mol
MDL Number: MFCD03844446
CAS Number: 99359-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR