Methyl-5-methylnicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methylnicotinate

A221580-100G A221580-1G A221580-5G
76721-984EA 256.74 USD
76721-984 76721-986 76721-988
Methyl-5-methylnicotinate 97%
Methyl-5-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD01075739
CAS Number: 29681-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR