Methyl 2-(quinolin-6-yl)acetate 98%
Supplier: Ambeed
|
Warning
|
A221429-100MG
A221429-10G
A221429-1G
A221429-250MG
A221429-5G
A221429-25G
76721-974EA
11
USD
76721-974
76721-976
76721-978
76721-980
76721-982
77879-351
Methyl 2-(quinolin-6-yl)acetate 98%
Methyl 2-(quinolin-6-yl)acetate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO₂ |
MDL Number:
MFCD04038669 CAS Number: 5622-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...