4-((4-Boronophenyl)amino)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

A221071-100MG A221071-1G A221071-250MG A221071-5G
76721-966EA 28.65 USD
76721-966 76721-968 76721-970 76721-972
4-((4-Boronophenyl)amino)-4-oxobutanoic acid 98%
4-((4-Boronophenyl)amino)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BNO₅
MW: 237.01 g/mol
MDL Number: MFCD03789260
CAS Number: 480424-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR