Methyl 4-methylthiazole-2-carboxylate 97%

Supplier: Ambeed
Warning

A221049-1G A221049-5G A221049-250MG
76721-962EA 19 USD
76721-962 76721-964 77879-350
Methyl 4-methylthiazole-2-carboxylate 97%
Methyl 4-methylthiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MDL Number: MFCD08275081
CAS Number: 14542-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR