(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate 95%

Supplier: Ambeed
Warning

A220803-25G A220803-5G
76721-958EA 39 USD
76721-958 76721-960
(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate 95%
(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₈
MDL Number: MFCD00799538
CAS Number: 167392-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR