2-(Naphthalen-2-yl)ethanol 95%

Supplier: Ambeed
Warning

A218419-1G A218419-250MG A218419-25G A218419-5G
76721-884EA 71 USD
76721-884 76721-886 76721-888 76721-890
2-(Naphthalen-2-yl)ethanol 95%
2-(Naphthalen-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MDL Number: MFCD00004128
CAS Number: 1485-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR