Milrinone 98%

Supplier: Ambeed
Danger

Synonyms: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile, 2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile, 3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone

A218047-100MG A218047-1G A218047-250MG A218047-25G A218047-5G
76721-874EA 8.49 USD
76721-874 76721-876 76721-878 76721-880 76721-882
Milrinone 98%
Milrinone

Precaución: for Research Use Only

Formula: C₁₂H₉N₃O
MW: 211.22 g/mol
MDL Number: MFCD00133539
CAS Number: 78415-72-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR