2-Phenyl-2-(2-pyridyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-2-(pyridin-2-yl)acetonitrile, 2-(alpha-Cyanobenzyl)pyridine, alpha-Phenyl-2-pyridineacetonitrile

A217898-1G A217898-5G
76721-870EA 66 USD
76721-870 76721-872
2-Phenyl-2-(2-pyridyl)acetonitrile 97%
2-Phenyl-2-(2-pyridyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
MDL Number: MFCD00023497
CAS Number: 5005-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR