Methyl 2-acetamido-2-(dimethoxyphosphoryl)acetate 97%

Supplier: Ambeed
Warning

A217016-100G A217016-10G A217016-1G A217016-250MG A217016-25G A217016-5G
76721-838EA 1676.23 USD
76721-838 76721-840 76721-842 76721-844 76721-846 76721-848
Methyl 2-acetamido-2-(dimethoxyphosphoryl)acetate 97%
Methyl 2-acetamido-2-(dimethoxyphosphoryl)acetate

Precaución: for Research Use Only

Formula: C₇H₁₄NO₆P
MW: 239.163 g/mol
MDL Number: MFCD08443722
CAS Number: 89524-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR