O-AC-L-Ser-OH 97%

Supplier: Ambeed
Warning

Synonyms: H-OAc-Ser-OH

A215243-250MG A215243-5G A215243-100MG
76721-822EA 65 USD
76721-822 76721-824 77879-322
O-AC-L-Ser-OH 97%
O-AC-L-Ser-OH

Precaución: for Research Use Only

Formula: C₅H₉NO₄
MW: 147.12 g/mol
MDL Number: MFCD00037771
CAS Number: 5147-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR