4-n-Pentylphenyl-4-n-propylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Pentylphenyl 4-propylbenzoate

A213738-10G A213738-1G A213738-25G A213738-5G
76721-802EA 20.16 USD
76721-802 76721-804 76721-806 76721-808
4-n-Pentylphenyl-4-n-propylbenzoate 97%
4-n-Pentylphenyl-4-n-propylbenzoate

Precaución: for Research Use Only

Formula: C₂₁H₂₆O₂
MW: 310.44 g/mol
MDL Number: MFCD00041941
CAS Number: 50649-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR