(S,Z)-5,6-Dihydroxy-4-((E)-3-(4-hydroxyphenyl)acryloyl)-2-(((S)-2,3,4-trihydroxy-5-((E)-3-(4-hydroxyphenyl)acryloyl)-6-oxo-3-((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)cyclohexa-1,4-dien-1-yl)methylene)-6-((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)cyclohex-4-ene-1,3-dione color value 30 (E1%,1cm,5)

Supplier: Ambeed
Warning

A213154-100G A213154-1G A213154-1MG A213154-250MG A213154-25G A213154-50MG A213154-5G
76721-778EA 1145.78 USD
76721-778 76721-780 76721-782 76721-784 76721-786 76721-788 76721-790
(S,Z)-5,6-Dihydroxy-4-((E)-3-(4-hydroxyphenyl)acryloyl)-2-(((S)-2,3,4-trihydroxy-5-((E)-3-(4-hydroxyphenyl)acryloyl)-6-oxo-3-((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)cyclohexa-1,4-dien-1-yl)methylene)-6-((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)cyclohex-4-ene-1,3-dione color value 30 (E1%,1cm,5)
(S,Z)-5,6-Dihydroxy-4-((E)-3-(4-hydroxyphenyl)acryloyl)-2-(((S)-2,3,4-trihydroxy-5-((E)-3-(4-hydroxyphenyl)acryloyl)-6-oxo-3-((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)cyclohexa-1,4-dien-1-yl)methylene)-6-((2R,3R,4S,5S,6

Precaución: for Research Use Only

Formula: C₄₃H₄₂O₂₂
MW: 910.78 g/mol
MDL Number: MFCD09954647
CAS Number: 36338-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR