2',4'-Dichloro-5'-hydroxyacetanilide 95%

Supplier: Ambeed
Danger

A212787-1G A212787-5G
76721-758EA 30 USD
76721-758 77568-052
2',4'-Dichloro-5'-hydroxyacetanilide 95%
2',4'-Dichloro-5'-hydroxyacetanilide

Precaución: For research use only.

Formula: C₈H₇Cl₂NO₂
MDL Number: MFCD00186236
CAS Number: 67669-19-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR