Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%

Supplier: Ambeed
Warning

A212441-250MG A212441-50MG A212441-100MG A212441-10MG A212441-1G
76721-748EA 219 USD
76721-748 76721-750 77600-712 77667-774 77667-776
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Precaución: for Research Use Only

Formula: C₃₂H₁₂BF₂₄K
MDL Number: MFCD00077934
CAS Number: 105560-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR