Methyl carbazate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate

A212052-100G A212052-25G
76721-738EA 12.74 USD
76721-738 77087-300
Methyl carbazate 97%
Methyl carbazate

Precaución: for Research Use Only

Formula: C₂H₆N₂O₂
MW: 90.08 g/mol
MDL Number: MFCD00007594
CAS Number: 6294-89-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR