4-Bromo-4'-chlorobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)(4-chlorophenyl)methanone

A211767-1G A211767-250MG A211767-5G
76721-728EA 31.83 USD
76721-728 76721-730 77600-700
4-Bromo-4'-chlorobenzophenone 98%
4-Bromo-4'-chlorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈BrClO
MW: 295.5590 g/mol
MDL Number: MFCD00045208
CAS Number: 27428-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR