Ethyl-4-hydroxy-8-methoxyquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxy-8-methoxyquinoline-3-carboxylate, Ethyl 1,4-dihydro-8-methoxy-4-oxoquinoline-3-carboxylate

A211696-10G A211696-1G A211696-25G A211696-5G
76721-712EA 26 USD
76721-712 76721-714 76721-716 76721-718 76721-720
Ethyl-4-hydroxy-8-methoxyquinoline-3-carboxylate 97%
Ethyl 4-hydroxy-8-methoxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₄
MDL Number: MFCD01829412
CAS Number: 27568-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR