N-Methylsaccharin 95%

Supplier: Ambeed
Warning

A211588-10G A211588-1G A211588-250MG A211588-25G A211588-5G
76721-696EA 121 USD
76721-696 76721-698 76721-700 76721-702 76721-704
N-Methylsaccharin 95%
N-Methylsaccharin

Precaución: for Research Use Only

Formula: C₈H₇NO₃S
MDL Number: MFCD00059263
CAS Number: 15448-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR