PP1 95%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butyl)-3-(p-tolyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

A211100-100MG A211100-10MG A211100-1MG A211100-50MG A211100-5MG
76721-686EA 497.57 USD
76721-686 76721-688 76721-690 76721-692 76721-694
PP1 95%
PP1

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₅
MW: 281.4 g/mol
MDL Number: MFCD01076570
CAS Number: 172889-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR