Chlorophyllin, coppered trisodium salt 98%
Supplier: Ambeed
|
Warning
|
A210676-100MG
A210676-25MG
A210676-5G
A210676-100G
A210676-25G
76721-676EA
6
USD
76721-676
76721-678
77600-696
77872-071
77872-072
Chlorophyllin, coppered trisodium salt 98%
Chlorophyllin, coppered trisodium salt
Precaución: for Research Use Only
Formula:
C₃₄H₃₁CuN₄Na₃O₆ MW: 724.15 g/mol |
MDL Number:
MFCD00012149 CAS Number: 11006-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...