Potassium (2,6-difluorophenyl)trifluoroboranuide 97%

Supplier: Ambeed
Warning

Synonyms: Potassium (2,6-difluorophenyl)trifluoroborate

A210219-1G A210219-25G A210219-5G
76721-664EA 13 USD
76721-664 76721-666 76721-668 76721-670
Potassium (2,6-difluorophenyl)trifluoroboranuide 97%
Potassium (2,6-difluorophenyl)trifluoroboranuide

Precaución: for Research Use Only

Formula: C₆H₃BF₅·K
MW: 219.989 g/mol
MDL Number: MFCD05865179
CAS Number: 267006-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR