Butyrylthiocholine iodide 98%

Supplier: Ambeed
Warning

Synonyms: S-n-Butyrylthiocholine iodide, (2-Butyrylthioethyl)trimethylammonium iodide, 2-(Butyrylthio)-N,N,N-trimethylethan-1-aminium iodide, (Propylcarbonylthioethyl)trimethylammonium iodide, (2-(Butyrylthio)ethyl)trimethylammonium iodide

A209743-10G A209743-25G A209743-5G A209743-1G
76721-658EA 41 USD
76721-658 76721-660 76721-662 77600-690
Butyrylthiocholine iodide 98%
Butyrylthiocholine iodide

Precaución: for Research Use Only

Formula: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
MDL Number: MFCD00011845
CAS Number: 1866-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR