2-Benzylaminoethanol 98%

Supplier: Ambeed
Warning

A208848-100G A208848-10G A208848-500G
76721-646EA 37 USD
76721-646 76721-648 76721-650
2-Benzylaminoethanol 98%
2-Benzylaminoethanol

Precaución: for Research Use Only

Formula: C₆H₅CH₂NHCH₂CH₂OH
MDL Number: MFCD00002840
CAS Number: 104-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR