4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
FAAH-IN-2, O-Desmorpholinopropyl Gefitinib, 4-(3-Chloro-4-fluoroanilino)-6-hydroxy-7-methoxyquinazoline, 4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-ol
A207764-1G
A207764-250MG
A207764-5G
A207764-100G
A207764-25G
A207764-1MG
76721-622EA
6.37
USD
76721-622
76721-624
77567-992
77600-678
77600-680
77667-744
4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol 98%
4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol
Precaución: For research use only.
Formula:
C₁₅H₁₁ClFN₃O₂ MW: 319.72 g/mol |
MDL Number:
MFCD09908046 CAS Number: 184475-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...