Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methoxycarbonylbenzoylacetate

A207510-1G A207510-250MG A207510-5G
76721-610EA 26.53 USD
76721-610 76721-612 76721-614
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate 98%
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₅
MW: 236.223 g/mol
MDL Number: MFCD00800637
CAS Number: 22027-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR