1-Boc-DL-nipecotic acid ethyl ester 95%

Supplier: Ambeed
Warning

A207234-100G A207234-10G A207234-5G
76721-600EA 25 USD
76721-600 76721-602 76721-604
1-Boc-DL-nipecotic acid ethyl ester 95%
1-Boc-DL-nipecotic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD04116274
CAS Number: 130250-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR