(Phenylthio)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Phenylthio)acetonitrile

A206811-1G A206811-25G A206811-5G
76721-582EA 22.28 USD
76721-582 76721-584 76721-586
(Phenylthio)acetonitrile 98%
(Phenylthio)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₇NS
MW: 149.22 g/mol
MDL Number: MFCD00019831
CAS Number: 5219-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR