N4-Acetyl-2'-deoxycytidine 97% (by HPLC)

Supplier: Ambeed
Warning

A205035-1G A205035-25G A205035-5G
76721-528EA 14.86 USD
76721-528 76721-530 76721-532
N4-Acetyl-2'-deoxycytidine 97% (by HPLC)
N4-Acetyl-2'-deoxycytidine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₅
MW: 269.25 g/mol
MDL Number: MFCD00057435
CAS Number: 32909-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR