Mucic acid 98%

Supplier: Ambeed
Warning

Synonyms: Galactaric acid, rel-(2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid, (2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid

A204823-100G A204823-25G A204823-5G A204823-1000G A204823-500G
76721-522EA 28.65 USD
76721-522 76721-524 76721-526 77667-714 77667-716
Mucic acid 98%
Mucic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₈
MW: 210.14 g/mol
MDL Number: MFCD00004239
CAS Number: 526-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR