Triethylmethylammonium bromide 99%

Supplier: Ambeed
Warning

Synonyms: N,N-Diethyl-N-methylethanaminium bromide, Triethylmethylamine hydrobromide

A202996-1G A202996-5G
76721-506EA 20 USD
76721-506 76721-508
Triethylmethylammonium bromide 99%
Triethylmethylammonium bromide

Precaución: for Research Use Only

Formula: C₇H₁₈BrN
MW: 196.13
MDL Number: MFCD00043177
CAS Number: 2700-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR