Sodium-2-ethylhexanoate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt

A200521-1000G A200521-100G A200521-500G
76721-434EA 64 USD
76721-434 76721-436 76721-438 76721-440
Sodium-2-ethylhexanoate 98%
Sodium-2-ethylhexanoate

Precaución: for Research Use Only

Formula: C₈H₁₅NaO₂
MDL Number: MFCD00014007
CAS Number: 19766-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR