N-Me-Ala-OtBu·HCl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 2-(methylamino)propanoate hydrochloride, N-Me-Ala-OtBu·HCl

A200288-10G A200288-1G A200288-250MG A200288-5G A200288-25G
76721-426EA 49 USD
76721-426 76721-428 76721-430 76721-432 77879-252
N-Me-Ala-OtBu·HCl 98%
N-Me-Ala-OtBu·HCl

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO₂
MW: 195.687 g/mol
MDL Number: MFCD08457605
CAS Number: 103614-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR