2-(Benzoylamino)benzoic acid 95%

Supplier: Ambeed
Warning

A200074-10G A200074-1G A200074-250MG A200074-5G
76721-418EA 249.32 USD
76721-418 76721-420 76721-422 76721-424
2-(Benzoylamino)benzoic acid 95%
2-(Benzoylamino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₃
MW: 241.2420 g/mol
MDL Number: MFCD00444173
CAS Number: 579-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR