3-Fluoro-N,N-dimethylaniline 97%

Supplier: Ambeed
Warning

A199409-100MG A199409-10G A199409-1G A199409-250MG A199409-5G
76721-402EA 33.95 USD
76721-402 76721-404 76721-406 76721-408 76721-410
3-Fluoro-N,N-dimethylaniline 97%
3-Fluoro-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD01862042
CAS Number: 2107-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR