O-Benzyl-DL-serine 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-OBzl-Ser-OH, H-DL-Ser(Bzl)-OH

A198677-10G A198677-1G A198677-25G A198677-5G
76721-366EA 116.7 USD
76721-366 76721-368 76721-370 76721-372
O-Benzyl-DL-serine 98%
O-Benzyl-DL-serine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
MDL Number: MFCD00021724
CAS Number: 5445-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR