N-Carbobenzoxy-DL-leucine 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: N-Cbz-DL-leucine, Z-DL-Leu-OH

A197359-1G A197359-25G A197359-5G
76721-338EA 6 USD
76721-338 76721-340 76721-342
N-Carbobenzoxy-DL-leucine 98% (by HPLC)
N-Carbobenzoxy-DL-leucine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD00065129
CAS Number: 3588-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR