N1,N2-Bis(2,6-diisopropylphenyl)ethane-1,2-diamine 97%

Supplier: Ambeed
Warning

A197072-10G A197072-1G A197072-25G A197072-5G
76721-330EA 172.93 USD
76721-330 76721-332 76721-334 76721-336
N1,N2-Bis(2,6-diisopropylphenyl)ethane-1,2-diamine 97%
N1,N2-Bis(2,6-diisopropylphenyl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₂₆H₄₀N₂
MW: 380.60 g/mol
MDL Number: MFCD10000903
CAS Number: 134030-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR