Z-Pyr-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Carbobenzoxy-5-oxopyrrolidine-2-carboxylic acid, N-Cbz-L-pyroglutamic acid, (S)-1-Benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, Z-Pyr-OH, N-Benzyloxycarbonyl-L-pyroglutamic acid, N-Carbobenzoxy-L-pyroglutamic acid, (S)-1-Cbz-5-oxopyrrolidine-2-carboxylic acid, (S)-1-((Benzyloxy)carbonyl)-5-oxopyrrolidine-2-carboxylic acid

A196655-10G A196655-25G A196655-5G
76721-324EA 13.8 USD
76721-324 76721-326 77600-608
Z-Pyr-OH 98%
Z-Pyr-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₅
MW: 263.249 g/mol
MDL Number: MFCD00037352
CAS Number: 32159-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR